missing translation for 'onlineSavingsMsg'
Learn More

Crolibulin, MedChemExpress

Product Code. 16485768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485768 10 mg 10mg
16458447 5 mg 5mg
16451927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485768 Supplier Medchem Express Supplier No. HY13603
 Restricted Item
Estimated Shipment: 30-11-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Crolibulin (EPC2407) is a tubulin polymerization inhibitor, with potent apoptosis induction and cell growth inhibition. Crolibulin has anti-tumor activity. Crolibulin also has cardiovascular toxicity and neurotoxicity.

Crolibulin (EPC2407) is a tubulin polymerization inhibitor, with potent apoptosis induction and cell growth inhibition. Crolibulin has anti-tumor activity. Crolibulin also has cardiovascular toxicity and neurotoxicity.

Spécification

Chemical Name or Material Crolibulin
CAS 1000852-17-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H17BrN4O3
Quantity 10 mg
Synonym EPC2407
Solubility Information DMSO : 125 mg/mL (299.57 mM; Need ultrasonic and warming)
SMILES N#CC1=C(N)OC2=C(N)C(N)=CC=C2[C@H]1C3=CC(OC)=C(OC)C(Br)=C3
Molecular Weight (g/mol) 417.26
Formula Weight 417.26
Percent Purity 98.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Afficher plus Afficher moins

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.