missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn sodium, MedChemExpress

Product Code. 16450878
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450878 5 g 5g
16422139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16450878 Supplier Medchem Express Supplier No. HYB0320A
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cromolyn sodium (Disodium Cromoglycate; FPL-670) is an antiallergic drug. Cromolyn sodium is a GSK-3β inhibitor with an IC50 of 2.0 μM.

Cromolyn sodium (Disodium Cromoglycate

Specifications

Chemical Name or Material Cromolyn sodium
CAS 15826-37-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H14Na2O11
Quantity 5 g
Synonym Disodium Cromoglycate FPL-670
Solubility Information H2O : 50 mg/mL (97.59 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (48.80 mM; Need ultrasonic)
SMILES OC(COC1=C2C(C=C(C(O[Na])=O)OC2=CC=C1)=O)COC3=C4C(C=C(C(O[Na])=O)OC4=CC=C3)=O
Molecular Weight (g/mol) 512.33
Formula Weight 512.33
Percent Purity 98.31%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.