missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148277
This item is not returnable.
View return policy
Description
CRP2 Polyclonal Antibody for Western Blot, IHC (P)
CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.
Specifications
| CRP2 | |
| Polyclonal | |
| Unconjugated | |
| CSRP2 | |
| AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 1466 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q16527 | |
| CSRP2 | |
| E. coli-derived Human CSRP2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction