missing translation for 'onlineSavingsMsg'
Learn More

CRYGB Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

198.00 € - 462.00 €

Specifications

Antigen CRYGB
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676861
View Documents
Novus Biologicals
NBP2-92172-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18687790
View Documents
Novus Biologicals
NBP2-92172-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

CRYGB Polyclonal antibody specifically detects CRYGB in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

CRYGB
Western Blot
Unconjugated
Rabbit
Mouse, Rat
CRYG2gamma-crystallin B, crystallin, gamma 1-2, crystallin, gamma B, gamma-B-crystallin, Gamma-crystallin 1-2
Recombinant fusion protein containing a sequence corresponding to amino acids 80-135 of human CRYGB (NP_005201.2). CLIPPHSGAYRMKIYDRDELRGQMSELTDDCISVQDRFHLTEIHSLNVLEGSWILY
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
1419
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.