missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSTF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85427-25ul
This item is not returnable.
View return policy
Description
CSTF3 Polyclonal antibody specifically detects CSTF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CSTF3 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CF-1 77 kDa subunit, Cleavage stimulation factor 77 kDa subunit, cleavage stimulation factor subunit 3, cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kD, cleavage stimulation factor, 3' pre-RNA, subunit 3, 77kDa, CSTF 77 kDa subunit, CSTF-77, MGC117398, MGC43001, MGC75122 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: QLWRDYNKYEEGINIHLAKKMIEDRSRDYMNARRVAKEYETVMKGLDRNAPSVPPQNTPQEAQQVDMWKKYIQWEKSNPLRTEDQTLITKRVMFAYEQCLLVLGHH | |
| 25 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 1479 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur