Learn More
CTGF/CCN2 Antibody (CL5339) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61415PCP
Product Code. 30288551
  • 737.00 € / 0.10mL
Description
CTGF/CCN2 Monoclonal antibody specifically detects CTGF/CCN2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTGF/CCN2
Monoclonal
PerCP
CCN2IGFBP-8, connective tissue growth factor, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
This antibody was developed using a recombinant protein derived from P29279, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer, Cell Biology, Cytokine Research, Growth and Development, Immunology, Innate Immunity
1490
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL5339
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified