missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTL1/SLC44A1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92231-0.02ml
This item is not returnable.
View return policy
Description
CTL1/SLC44A1 Polyclonal antibody specifically detects CTL1/SLC44A1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CTL1/SLC44A1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| CD92 antigen, CDw92, CDW92 antigen, CDW92choline transporter-like protein 1, CHTL1, CTL1CD92RP11-287A8.1, Solute carrier family 44 member 1, solute carrier family 44, member 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 55-210 of human SLC44A1 (NP_536856.2). AARLVSGYDSYGNICGQKNTKLEAIPNSGMDHTQRKYVFFLDPCNLDLINRKIKSVALCVAACPRQELKTLSDVQKFAEINGSALCSYNLKPSEYTTSPKSSVLCPKLPVPASAPIPFFHRCAPVNISCYAKFAEALITFVSDNSVLHRLISGVMT | |
| 0.02 mL | |
| Cancer, CD Markers, Endocrinology, Immunology, Signal Transduction | |
| 23446 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction