missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CTLA-4 Antibody (ipilimumab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28243-50ug
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CTLA-4 | |
| Monoclonal | |
| Unconjugated | |
| P16410 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ipilimumab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| CTLA-4/ CD152 | |
| 50 μg | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Immune System Diseases, Immunology, Neuroscience, Signal Transduction | |
| 1493 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction