missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTLA-4 Monoclonal Antibody (WKH 203), PE

Mouse Monoclonal Antibody
277.00 € - 614.00 €
Specifications
| Antigen | CTLA-4 |
|---|---|
| Clone | WKH 203 |
| Concentration | 0.1 mg/mL |
| Content And Storage | 4°C, do not freeze |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15348490
![]() |
Invitrogen™
MA517586 |
50 μg |
277.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15358490
|
Invitrogen™
MA517587 |
200 μg |
614.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CTLA-4 Monoclonal Antibody for Flow
CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.Specifications
| CTLA-4 | |
| 0.1 mg/mL | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| 0 | |
| 63835 | |
| Purified rCTLA-4hlg fusion protein. | |
| Antibody |
| WKH 203 | |
| 4°C, do not freeze | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Rat | |
| CTLA4 | |
| ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form | |
| CTLA4 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title