missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTRP3 Monoclonal Antibody (860101)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561885
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.
Specifications
| CTRP3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q9BXJ4 | |
| C1qtnf3 | |
| Mouse myeloma cell line, NS0-derived human CTRP3/C1qTNF3/CORS26, Gln23-Lys246. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| ELISA | |
| 860101 | |
| Unconjugated | |
| C1qtnf3 | |
| 2310005P21Rik; C1ATNF3; C1q and tumor necrosis factor related protein 3; C1qtnf3; cartducin; cartonectin; collagenous repeat-containing sequence 26 kDa protein; collagenous repeat-containing sequence of 26-kDa; complement C1q tumor necrosis factor-related protein 3; Corcs; CORS; CORS26; CORS-26; CTRP3; Secretory protein CORS26; UNQ753/PRO1484 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 114899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction