missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTRP3 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148594

Product Code. 30536049

  • 499.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTRP3
Polyclonal
Unconjugated
C1qtnf3
2310005P21Rik; C1ATNF3; C1q and tumor necrosis factor related protein 3; C1qtnf3; cartducin; cartonectin; collagenous repeat-containing sequence 26 kDa protein; collagenous repeat-containing sequence of 26-kDa; complement C1q tumor necrosis factor-related protein 3; Corcs; CORS; CORS26; CORS-26; CTRP3; Secretory protein CORS26; UNQ753/PRO1484
Goat
Antigen affinity chromatography
RUO
81799
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9ES30
C1qtnf3
Mouse myeloma cell line NS0-derived recombinant mouse CTRP3/C1qTNF3/CORS26, Gln23-Lys246, predicted.
100 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.