missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP7 Antibody (K14-S), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-30132-0.05ML
This item is not returnable.
View return policy
Description
CTRP7 Monoclonal specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| CTRP7 | |
| Monoclonal | |
| 3.6mg/mL | |
| Western Blot 1:5000, ELISA 1:20000-1:50000 | |
| Q9BXJ2 | |
| C1QTNF7 | |
| Peptide from near the carboxy terminus of human CTRP7. | |
| 0.05 mL | |
| Primary | |
| This antibody is specific to the carboxy terminus of CTRP7 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| K14-S | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
| C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 114905 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction