missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ One SFM

Product Code. 30283674 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 L
Format:
BPC
Bottle
Unit Size:
1 L
1000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Format Unit Size
30283674 1 L BPC 1 L
30282486 1 L Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30283674 Supplier Gibco™ Supplier No. A5757202
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies.

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies. This medium improves cell count and early memory phenotype throughout all stages of expansion.

Benefits:

  • Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
  • Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
  • Expands both healthy and disease state donors
  • Supports efficient viral and non-viral editing with downstream expansion
  • Mitigates risk of contaminants and variability associated with animal-sourced materials
  • Supports scale-up via larger catalog pack sizes

Features:

  • Animal-origin-free (AOF) formulation
  • Serum-free—designed without the need for serum or serum replacement
  • No separate components—no need to mix basal medium with supplement to obtain complete medium
  • Yields high density cultures
  • Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD28 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
  • Available in 1 L bottle, 1 L bag, 5 L bag, and 10 L bag options

Increased efficiency and consistent performance:

CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.

Simplify your transition into clinical research:

The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)
TRUSTED_SUSTAINABILITY

Specifications

Cell Type T cells
Classification Animal Origin-free, Serum-free, cGMP
Form Liquid
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Glutamine, No Phenol Red
Product Line CTS OpTmizer
Quantity 1 L
Format BPC
Content And Storage Store at 2-8°C and protect from light.
Culture Type Suspension Cell Culture, Dynamic Bioreactors
Show More Show Less
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Does CTS OpTmizer One SFM work in bioreactors?

Yes. CTS OpTmizer One SFM offers excellent performance in a range of culture vessels, from static to dynamic bioreactors.

Do I need to add serum to CTS OpTmizer One SFM?

No. CTS OpTmizer One SFM was designed to be used without serum or serum replacement supplementation.

Does CTS OpTmizer One SFM contain protein?

Yes, all protein within CTS OpTmizer One SFM is recombinant.

Does CTS OpTmizer One SFM contain L-glutamine?

No. The most consistent and reproducible results are seen when L-glutamine is added to CTS OpTmizer One SFM at the point of use.

Is CTS OpTmizer One SFM an animal-origin-free (AOF) version of legacy CTS OpTmizer SFM?

No. CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium. It is not a kit like the legacy CTS OpTmizer SFM, and does not require mixing of separate basal medium and supplement components to create complete medium.

CTS OpTmizer One SFM does not contain or use in the manufacturing processes any primary or secondary raw materials that are derived directly from animal (including human) tissue or body fluid.


For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.