missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ Pro SFM, no phenol red

Product Code. 30284086 Shop All Gibco Products
Change view
Click to view available options
Quantity:
5 L
10 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30284086 10 L Each
30282802 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30284086 Supplier Gibco™ Supplier No. A4966107
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies.

Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies. This medium improves central memory phenotype and cell growth by shifting the cell's metabolism. The result is a larger population of central memory cells in a shorter period of time.

CTS OpTmizer Pro SFM consists of CTS OpTmizer Pro Basal Medium and CTS OpTmizer Supplement, which are mixed together prior to use.

CTS OpTmizer Pro SFM in 5 L and 10 L bag format is compatible with closed-system processing for media preparation and integration to standard cell therapy equipment, including CTS devices (Xenon, Rotea, and DynaCellect).

Benefits of CTS OpTmizer Pro SFM include:

  • Supplement component now in bag format to allow for closed mixing with basal medium
  • Reduces the number of open steps in your process, thus reducing contamination risks
  • Easy integration with cell therapy equipment, including CTS Xenon, Rotea, and DynaCellect
  • Maintains central memory T cells further into expansion and is well suited for longer workflows (>15 days)
  • Offers >90% (average) T-cell fold-expansion (from healthy donors) compared to CTS OpTmizer medium
  • Modest increases in central memory cells at early stage in expansion protocol result in larger cell yields at harvest
  • Helps achieve the same number of cells in a shorter duration
  • Ideally suited to the production of allogeneic therapies

Features include:

  • Flexible pack sizes in Aegis5-14 film: basal medium in 5 L and 10 L bags
  • Weldable C-Flex and PVC lines
  • Supports allogeneic T-cell processes through all stages of development and manufacturing
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols

Increased efficiency and consistence performance
CTS OpTmizer Pro SFM, no phenol red, is serum-free and xeno-free and supports high density cultures in static cultures. It displays consistent performance in the support of T-cell expansion with better central memory phenotype in a shorter time.

Enhanced expansion
CTS OpTmizer Pro SFM, no phenol red, increases the number of central memory cells at an early stage in the expansion process and maintains T-cell functionality.

Enhanced design and compatibility
The supplement component is now in a bag with a weldable PVC line, allowing for closed-system mixing with the CTS OpTmizer Pro basal medium bag. This feature, along with larger 5 L and 10 L basal medium sizes, addresses manufacturers' need for scale, efficiency, and contamination risk mitigation.

Use of Thermo Scientific™ Aegis5-14 Film allows compatibility with closed systems. This film is a five-layer cast film produced in a cGMP facility. The outer layer is a polyester elastomer coextruded with an ethyl vinyl alcohol (EVOH) barrier layer and an ultra-low density polyethylene product contact layer. Aegis5-14 Film is manufactured using no animal derived components.

Simplify your transition into clinical research
The Gibco™ Cell Therapy Systems (CTS) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices.

CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)
TRUSTED_SUSTAINABILITY

Specifications

Classification Xeno-free
Form Liquid
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Phenol Red
Product Line OpTmizer™ Pro
Quantity 10 L
Shipping Condition Room Temperature
Culture Type Static Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.