missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ OpTmizer™ T-Cell Expansion SFM, no phenol red, bottle format

Product Code. 15949695 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15949695 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15949695 Supplier Gibco™ Supplier No. A3705001
Make Your Process Development Count
Use promo code 27978
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gibco CTS OpTmizer T-Cell Expansion SFM supports the growth and expansion of human T lymphocytes.

CTS OpTmizer T-Cell Expansion SFM is available with and without phenol red. This version of CTS OpTmizer SFM lacks phenol red, and is available in both bottle and bag format.

Features of CTS OpTmizer T-Cell Expansion SFM include:
• Supports high-density T cell culture (>4 x 106 CD3+ T-cells/mL)
• Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols
• Similar phenotype, function (e.g., cytokine secretion profile), and viability to T-cells cultured with conventional human AB serum-supplemented medium
• Supports a T-cell phenotype similar to human serum-supplemented medium
• Demonstrates enhanced efficacy and persistence of CART-19 cells when grown in medium supplemented with CTS Immune Cell Serum Replacement (ICSR)

Demonstrated functional performance
Cells grown in CTS OpTmizer T-Cell Expansion SFM with or without human serum demonstrate good cytokine production profiles in polyclonal activated T cells. CTS OpTmizer T-Cell Expansion SFM maintains the desired the CD62L+ central memory T-cell phenotype.

GMP manufactured
The primary manufacturing site for CTS OpTmizer T-Cell Expansion SFM is FDA-registered and has an ISO13485-certified quality management system. The methods and controls used for the manufacturing are in conformity with current Good Manufacturing Practices for medical devices, 21 CFR Part 820, of the regulation. Alternate manufacturing sites are available, providing risk mitigation by helping to ensure security of supply.

Versatility
CTS OpTmizer T-Cell Expansion SFM has been widely adopted in clinical T-cell research applications including the culture of CAR T-cells. It works with multiple culture formats, including culture flasks, culture bags, and rocking bioreactor systems. CTS OpTmizer T-Cell Expansion SFM was designed to support expansion of cultured human T-cells without additional serum supplementation. Increased performance is generally observed when supplemented with CTS Immune Cell Serum Replacement.

What you get
CTS OpTmizer T-Cell Expansion SFM includes two components: 1 liter of CTS OpTmizer T-Cell Expansion Basal Medium and 26 mLs of CTS OpTmizer T-Cell Expansion Auxiliary Concentrated Medium, which are mixed together to create the complete medium. Full traceability documentation such as Certificate of Origin and Certificate of Analysis are available.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells
Product Type Serum Free Medium (SFM)
Without Additives No Phenol Red
Product Line OpTmizer
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage • 1 x 1000 mL bottle CTS OpTmizer Expansion Basal Medium, store at 2°C to 8°C, protect from light
• 1 x 26 mL CTS OpTmizer Expansion Supplement, store in the dark (at 2°C to 8°C.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.