missing translation for 'onlineSavingsMsg'
Learn More

Cumene, 99%, pure

Product Code. 10417380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10417380 1 L Glass bottle 1 L
10114300 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10417380 Supplier Thermo Scientific Acros Supplier No. 181010010
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-82-8 | C9H12 | 120.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.19
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96.0°C
Color Colorless
Density 0.8620g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)2
Refractive Index 1.4905 to 1.4925
Quantity 1 L
Beilstein 05, 393
Merck Index 15, 2607
Specific Gravity 0.862
Solubility Information Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone
Viscosity 0.79 mPa.s (20°C)
Formula Weight 120.19
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material Cumene, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.