missing translation for 'onlineSavingsMsg'
Learn More

Cumene, 99%

Product Code. 11443834
Change view
Click to view available options
Quantity:
250 mL
500 mL
2500 mL
Unit Size:
2500mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443834 2500 mL 2500mL
11394539 250 mL 250mL
11433834 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443834 Supplier Thermo Scientific Alfa Aesar Supplier No. A11864.0F
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-82-8 | C9H12 | 120.195 g/mol

Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cumene is used in the production of phenol and acetone. It is used as a thinner for paints, lacquers, and enamels and as a component of high octane fuels. It is used in the manufacture of phenol, acetone, acetophenone, and methylstyrene.

Solubility
Soluble in alcohol, ether, acetone, benzene, carbon tetrachloride. Insoluble in water.

Notes
Store at 2-8°C. Avoid prolonged storage with exposure to air. Incompatible with acids, oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-82-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008881
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
Synonym isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Melting Point -96°C
Density 0.864
Boiling Point 152°C to 154°C
Flash Point 46°C (114°F)
Assay Percent Range 99%
Refractive Index 1.491
Quantity 2500 mL
UN Number UN1918
Beilstein 1236613
Merck Index 14,2617
Solubility Information Soluble in alcohol,ether,acetone,benzene,carbon tetrachloride. Insoluble in water.
Formula Weight 120.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cumene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.