missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Melting Point | -30.0°C |
| Density | 1.0600g/mL |
| Flash Point | 83°C |
| Assay Percent Range | 80 to 85% (Iodometry) |
| Packaging | Plastic bottle |
| Linear Formula | C6H5C(CH3)2OOH |
| Quantity | 250 g |
| Specific Gravity | 1.06 |
| Solubility Information | Solubility in water: miscible. |
| IUPAC Name | 2-hydroperoxypropan-2-ylbenzene |
| Show More |
Safety and Handling
Product Identifier
- Cumyl hydroperoxide
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Organic peroxide Type E & F
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H242-Heating may cause a fire.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Cumene hydroperoxide, Isopropylbenzene,Benzenemethanol, Acetophenone
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?