missing translation for 'onlineSavingsMsg'
Learn More

Cumyl hydroperoxide, tech. 80%

Product Code. 11347026
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347026 500 g 500g
11337026 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347026 Supplier Thermo Scientific Alfa Aesar Supplier No. L06866.36
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-15-9 | C9H12O2 | 152.19 g/mol

Cumene hydroperoxide is used in the preparation of polystyrene nanocapsules. It acts as a curing agent for polyester resins and as an oxidizer in organic chemical reactions. It serves as an initiator for radical polymerization especially for acrylate and methacrylate monomers. It also employed as an intermediate in the cumene process for developing phenol and acetone from benzene and propene. Further, it is used as an epoxidation reagent for allylic alcohols and fatty acid esters. In addition to this, it is also used to prepare methylstyrene, acetophenone and cumyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cumene hydroperoxide is used in the preparation of polystyrene nanocapsules. It acts as a curing agent for polyester resins and as an oxidizer in organic chemical reactions. It serves as an initiator for radical polymerization especially for acrylate and methacrylate monomers. It also employed as an intermediate in the cumene process for developing phenol and acetone from benzene and propene. Further, it is used as an epoxidation reagent for allylic alcohols and fatty acid esters. In addition to this, it is also used to prepare methylstyrene, acetophenone and cumyl alcohol.

Solubility
Miscible with alcohol, acetone, ether, esters, hydrocarbons and chlorinated hydrocarbons. Slightly miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with powdered metals, organic materials, heavy metal salts, metal salts, combustible materials, acids, alkalis, reducing agents, rust, charcoal, amines, copper, lead, cobalt and cobalt oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-15-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002129
InChI Key YQHLDYVWEZKEOX-UHFFFAOYSA-N
Synonym cumene hydroperoxide, cumyl hydroperoxide, hydroperoxide, 1-methyl-1-phenylethyl, cumenyl hydroperoxide, alpha,alpha-dimethylbenzyl hydroperoxide, cumolhydroperoxid, 7-cumyl hydroperoxide, cumolhydroperoxide, cument hydroperoxide, hydroperoxyde de cumene
PubChem CID 6629
ChEBI CHEBI:78673
IUPAC Name 2-hydroperoxypropan-2-ylbenzene
SMILES CC(C)(OO)C1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.03
Boiling Point 100°C to 102°C (8 mmHg)
Flash Point 56°C (132°F)
Refractive Index 1.524
Quantity 500 g
UN Number UN3109
Beilstein 1908117
Solubility Information Miscible with alcohol,acetone,ether,esters,hydrocarbons and chlorinated hydrocarbons. Slightly miscible with water.
Formula Weight 152.2
Percent Purity 80%
Grade Technical
Chemical Name or Material Cumene hydroperoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.