missing translation for 'onlineSavingsMsg'
Learn More

Cupferron, 97+%

Product Code. 15483137
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483137 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15483137 Supplier Thermo Scientific Alfa Aesar Supplier No. A16551.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 135-20-6 | C6H9N3O2 | 155.16 g/mol

Reagent for determination of Ce, Cu, Fe, Sn, and Ti

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 135-20-6
Melting Point 154°C (decomposition)
Odor Odorless
Molecular Formula C6H9N3O2
MDL Number MFCD00078422
Quantity 25 g
UN Number UN2811
Merck Index 14,2622
Synonym cupferron, copperone, azanyliumylidene n-nitroso-n-oxidoaniline
InChI Key NJVHCUNZAMFQNA-UHFFFAOYSA-N
SMILES N.ON(N=O)C1=CC=CC=C1
IUPAC Name azanium;N-oxido-N-phenylnitrous amide
Molecular Weight (g/mol) 155.16
PubChem CID 2724103
Formula Weight 155.16
Percent Purity ≥97%
Chemical Name or Material Cupferron
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.