missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Curcumin

Product Code. 18187963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18187963 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18187963 Supplier R&D Systems™ Supplier No. NBP2262435G
  • ONLINE EXCLUSIVE PRICE
    376.50 €
    308.70 € / 5g
    Save 67.80 €
    18% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. For use in research applications.

  • Linear Formula: [HOC6H3(OCH3)CH=CHCO]2CH2
  • Molecular Weight: 368.38
  • Beilstein Registry Number: 2306965
  • Colour Index Number: 75300
  • EC Number:207-280-5
  • MDL number: MFCD00008365
  • PubChem ID: 24892408

Reconstitution: Reconstitute with DMSO to bring curcumin to a final concentration of 11 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

CAS 458-37-7
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Line Novus
Quantity 5 g
Species Human
Target Curcumin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.