missing translation for 'onlineSavingsMsg'
Learn More

Curcumin, TRC

Product Code. 17981045
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981045 50 mg 50mg
17964855 1 g 1g
17982075 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981045 Supplier Toronto Research Chemicals Supplier No. TRCC83850050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458-37-7
Molecular Formula C21 H20 O6
Molecular Weight (g/mol) 368.38
Synonym 1,6-Heptadiene-3,5-dione, 1,7-bis(4-hydroxy-3-methoxyphenyl)-, (E,E)- (8CI), Curcumin (6CI), (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, (E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, (E,E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, BR 213, Biocurcumax, C Yellow 15, C.I. 75300, C.I. Natural Yellow 3, Colorant E 100, Curcuma, Curcumax, Curcumin I, Curcumin W, Curcumine, Cureit, Diferuloylmethane, E 100, E 100 (dye), Haidr, Halad, Haldar, Halud, INS 100i, Indian Saffron, Jianghuangsu, Kacha Haldi, Kiriyasu Oil Yellow U, Lipocurc, Longvida, Merita Earth, NSC 32982, NanoCurc, Natural Yellow 3, Natxtra, San-Ei Curcumine AL, San-Ei Gen Curcumine AL, Souchet, Terra Merita, Theracurmin, Theracurmin CR 011L, Turmeric, Turmeric (dye), Turmeric yellow, Ukon, Ukon (dye), Vacurin, Yellow Ginger, Yellow Root, Yo-Kin, trans,trans-Curcumin
IUPAC Name (1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
SMILES COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+
Formula Weight 368.126
Percent Purity >95%
Chemical Name or Material Curcumin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.