missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4390829
Additional Details : Weight : 0.00450kg
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C | |
siRNA | |
Ambion™, Silencer™ Select | |
Standard |
Custom Select siRNA, 40nmol, Standard Purity, ≥80% Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
Custom Select siRNA | |
40 nmol |
For Research Use Only. Not for use in diagnostic procedures.