Learn More
Invitrogen™ Custom Select siRNA
Product Code. 15571084
Change view
Click to view available options
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Unit Size:
20nmol
250nmol
40nmol
5nmol
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15571084 1 μmol Each
15531044 5 nmol 5nmol
15551044 40 nmol 40nmol
15541044 20 nmol 20nmol
15561044 20 nmol 20nmol
15571044 40 nmol 40nmol
15561064 250 nmol 250nmol
15561084 10 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15571084 Supplier Invitrogen™ Supplier No. 4444245
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Custom Select siRNA
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C
RNAi Type siRNA
Product Line Silencer Select, Ambion
Product Type siRNA
Purity HPLC
Quantity 1 μmol