missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Select siRNA, In Vivo Ready

Product Code. 15511064
Change view
Click to view available options
Quantity:
1 μmol
250 nmol
Unit Size:
250nmol
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15511064 250 nmol 250nmol
15521064 1 μmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15511064 Supplier Ambion™ Supplier No. 4404006
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Custom Select siRNA, In Vivo Ready

Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description In-vivo Ready Custom Select siRNA, 250nmol, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities
For Use With (Application) Knocking down gene expression in order to study protein function in a wide range of cell types
Product Type In-vivo Ready Custom Select siRNA
Quantity 250 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.