Learn More
Custom Select siRNA, In Vivo Ready
Product Code. 15531064
Change view
Click to view available options
Quantity:
10 μmol
Unit Size:
Each
Product Code. Quantity Unit Size
15531064 10 μmol Each
1 options
Product Code. 15531064 Supplier Invitrogen Supplier No. 4404008
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Custom Select siRNA, In Vivo Ready
  • Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
  • These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C
RNAi Type siRNA
Product Line Silencer Select, Ambion
Product Type siRNA
Quantity 10 μmol