missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Silencer™ siRNA

Product Code. 15591144
Change view
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15591144 20 nmol 20nmol
15541144 20 nmol 20nmol
15551144 40 nmol 40nmol
15561144 40 nmol 40nmol
15571144 20 nmol 20nmol
15581144 20 nmol 20nmol
15501154 100 nmol 100nmol
15521154 1 umol 1µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15591144 Supplier Ambion™ Supplier No. AM16212
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom Silencer™ siRNA

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description Custom siRNA, 20nmol, FAM Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
For Use With (Application) RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
Product Type Custom siRNA
Purity FAM
Quantity 20 nmol
Where should I add a fluorescent tag to minimize any negative functional effect on my siRNA?

For any RNAi duplex (miRNA or siRNA) there is a possibility that adding a modification will affect how the antisense strand interacts with RISC and aligns with its mRNA target, thus impacting functionality. For that reason, we recommend that you label the 5' end of the sense strand for siRNA (or passenger strand for miRNA). Since the 5' end of the antisense (guide strand) is what gets recognized by RISC, it is best to add the fluorescent modification to the end of the duplex on the strand that does not interact with RISC. You can also test a labeled vs. non-labeled RNAi duplex to determine if the modification is affecting the siRNA or miRNA functionality.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.