missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Silencer™ siRNA, Standard Purity

Product Code. 15501064
Change view
Click to view available options
Quantity:
5 nmol
Unit Size:
5nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15501064 5 nmol 5nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15501064 Supplier Ambion™ Supplier No. 4404004
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom Silencer™ siRNA, Standard Purity

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water
Description Custom siRNA, 5nmol, Standard Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
RNAi Type siRNA
For Use With (Application) RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
Product Line Silencer, Ambion
Product Type Custom siRNA
Purity Standard
Quantity 5 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.