missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CXCL12/SDF-1 Antibody (Genentech anti-CXCL12) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28766-100ug
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CXCL12/SDF-1 | |
| Monoclonal | |
| Unconjugated | |
| P48061 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech anti-CXCL12 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
| CXCL12/ SDF1a | |
| 100 μg | |
| Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers | |
| 6387 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction