Learn More
Novus Biologicals™ CXCL12/SDF-1 Antibody (Genentech anti-CXCL12) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28766-50ug
 View more versions of this product
Product Code. 30048107
  • 213.00 € / 50µg
Description
CXCL12/SDF-1 Monoclonal antibody specifically detects CXCL12/SDF-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL12/SDF-1
Monoclonal
Unconjugated
P48061
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Genentech anti-CXCL12
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
CXCL12/ SDF1a
50 μg
Chemokines and Cytokines, Growth and Development, Neuronal Cell Markers
6387
Human
Purified