Learn More
CXCL13/BLC/BCA-1 Antibody (CL10543) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-07988AF594
Product Code. 30290707
  • 737.00 € / 0.10mL
Description
CXCL13/BLC/BCA-1 Monoclonal antibody specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL13/BLC/BCA-1
Monoclonal
Alexa Fluor 594
ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
This antibody was developed using a synthetic peptide derived from O43927, with the exact immunogen sequence remaining proprietary.
0.1 mL
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction
10563
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10543
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified