missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL13/BLC/BCA-1 Antibody (MM0212-12A2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
218.00 € - 533.00 €
Specifications
| Antigen | CXCL13/BLC/BCA-1 |
|---|---|
| Clone | MM0212-12A2 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18021417
|
Novus Biologicals
NBP2-12222-0.025MG |
0.025 mg |
218.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758313
|
Novus Biologicals
NBP2-12222 |
533.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
| CXCL13/BLC/BCA-1 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| CXCL13 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0212-12A2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| O43927 | |
| 10563 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title