Learn More
Invitrogen™ CXCL5 Recombinant Rabbit Monoclonal Antibody (9H45L5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 700656
Description
Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
Specifications
| CXCL5 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P42830 | |
| CXCL5 | |
| human CXCL5 protein corresponding to amino acids 37-114. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 9H45L5 | |
| Unconjugated | |
| CXCL5 | |
| AMCF-II; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6374 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.