missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL8/IL-8 Antibody (SR1081), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22423-50ul
This item is not returnable.
View return policy
Description
CXCL8/IL-8 Monoclonal antibody specifically detects CXCL8/IL-8 in Human samples. It is validated for Western Blot
Specifications
| CXCL8/IL-8 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1081 | |
| Western Blot 1:500-1:2000 | |
| 3-10C, AMCF-I, C-X-C motif chemokine 8, CXCL8SCYB8, Emoctakin, GCP-1TSG-1, interleukin 8, K60, LECT, MDNCFb-ENAP, member 8, MONAPGCP1, NAP-1NAP1, Neutrophil-activating protein 1, Protein 3-10C, T cell chemotactic factor, T-cell chemotactic factor | |
| A synthesized peptide derived from human CXCL8/IL-8 (Uniprot #: P10145) | |
| 50 μg | |
| Angiogenesis, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology, Innate Immunity, Signal Transduction | |
| 3576 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction