missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL9/MIG Rabbit anti-Mouse, Clone: 277, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90453-50UL
This item is not returnable.
View return policy
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Mouse samples. It is validated for ELISA.
Specifications
| CXCL9/MIG | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| 277 | |
| ELISA 1:5000-1:10000 | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| Recombinant Mouse CXCL9/MIG protein | |
| 50 μg | |
| Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction | |
| 4283 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction