missing translation for 'onlineSavingsMsg'
Learn More

α-Cyano-βphenylacrylic Acid, MP Biomedicals

Product Code. 11443125 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443125 Supplier MP Biomedicals Supplier No. 0521374005
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 1011-92-3
Molecular Formula C10H7NO2
Quantity 5 g
Synonym 2-cyano-3-phenylacrylic acid, alpha-cyanocinnamicacid, cyanocinnamic acid, 2-cyano-3-phenyl-2-propenoic acid, acmc-1b4v0, a-cyano-b-phenylacrylic acid, alpha-cyano-beta-phenyl-acrylic acid
InChI Key CDUQMGQIHYISOP-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C=C(C#N)C(=O)O
IUPAC Name 2-cyano-3-phenylprop-2-enoic acid
Molecular Weight (g/mol) 173.171
PubChem CID 70534
Chemical Name or Material α-Cyano-β-Phenylacrylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.