missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYBB/NOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
218.00 € - 617.00 €
Specifications
| Antigen | CYBB/NOX2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613429
|
Novus Biologicals
NBP2-41291-0.025mg |
0.025 mg |
218.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701784
|
Novus Biologicals
NBP2-41291 |
0.1 mg |
617.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYBB/NOX2 Polyclonal specifically detects CYBB/NOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| CYBB/NOX2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 1536 | |
| Antibody was raised against a 15 amino acid peptide near the amino terminus of human NOX2. The immunogen is located within amino acids 140 - 190 of NOX2. Amino Acid Squence: NFARKRIKNPEGGLY | |
| Primary | |
| At least two isoforms are known to exist; this antibody will detect both isoforms. NOX2 is predicted to not cross-react with other NOX proteins. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit | |
| CYBB | |
| IgG | |
| Affinity Purified | |
| 58 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title