missing translation for 'onlineSavingsMsg'
Learn More

Cyclamic acid, MedChemExpress

Product Code. 16492219
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492219 1 g 1g
16420988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492219 Supplier Medchem Express Supplier No. HYB0541
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyclamic acid (Cyclohexylsulfamic acid) is one of the most widely used artificial sweetenersin food and pharmaceuticals.

Cyclamic acid (Cyclohexylsulfamic acid) is one of the most widely used artificial sweetenersin food and pharmaceuticals.

Specifications

Chemical Name or Material Cyclamic acid
Physical Form Solid
CAS 100-88-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C6H13NO3S
Synonym Cyclohexylsulfamic acid Cyclamate
Solubility Information DMSO : 100 mg/mL (557.91 mM; Need ultrasonic)
SMILES O=S(O)(NC1CCCCC1)=O
Molecular Weight (g/mol) 179.24
Formula Weight 179.24
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.