missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin A1/Cyclin A2 Antibody (SR1909), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | Cyclin A1/Cyclin A2 |
|---|---|
| Clone | SR1909 |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670204
|
Novus Biologicals
NBP3-21690-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625474
|
Novus Biologicals
NBP3-21690-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin A1/Cyclin A2 Monoclonal antibody specifically detects Cyclin A1/Cyclin A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| Cyclin A1/Cyclin A2 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| IgG | |
| Affinity purified |
| SR1909 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| A synthesized peptide derived from human Cyclin A1/Cyclin A2 (Uniprot #: P20248, P78396) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title