missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (CL12328), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21214-25ul
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human samples. It is validated for Immunofluorescence
Specifications
| Cyclin D1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| CL12328 | |
| Immunocytochemistry/Immunofluorescence 2-10 ug/ml | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| This antibody was developed against a Synthetic Peptide of P24385. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| 595 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction