missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558015
This item is not returnable.
View return policy
Description
Immunogen sequence: LLGAVCMFL ASKLKETSPL TAEKLCIYTD NSIKPQELLE WELVVLGKLK WNLAAVTPHD FIEHILRKLP QQREKLSLIR KHAQTFIALC ATDFKFAMYP PS.
Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
Specifications
| Cyclin D2 | |
| Recombinant Monoclonal | |
| 0.63 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P30279 | |
| CCND2 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Cyclin D2 (NP_001750.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 5B4M7 | |
| Unconjugated | |
| CCND2 | |
| 2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene | |
| Rabbit | |
| Protein A | |
| RUO | |
| 894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction