missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cyclin D2 Recombinant Rabbit Monoclonal Antibody (5B4M7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558016

 View more versions of this product

Product Code. 30284714

  • 436.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LLGAVCMFL ASKLKETSPL TAEKLCIYTD NSIKPQELLE WELVVLGKLK WNLAAVTPHD FIEHILRKLP QQREKLSLIR KHAQTFIALC ATDFKFAMYP PS.

Cyclin D2 is a G1 cyclin required for G1 phase progression and is a strong candidate for a proto-oncogene. Cyclin D2 can phosphorylate pRB when associated with cdk 4 and / or cdk 6.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin D2
Recombinant Monoclonal
0.63 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P30279
CCND2
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Cyclin D2 (NP_001750.1).
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
5B4M7
Unconjugated
CCND2
2600016F06Rik; AI256817; BF642806; C86853; CCND2; cD2; cyclin D2; Cyl-2; G1/S-specific cyclin-D2; KIAK0002; MGC102758; MPPH3; Vin1; Vin-1; Vin-1 proto-oncogene
Rabbit
Protein A
RUO
894
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.