missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine hydrochloride, MedChemExpress

Product Code. 16451048
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451048 5 g 5g
16432269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451048 Supplier Medchem Express Supplier No. HYB0740
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyclobenzaprine hydrochloride (MK130 hydrochloride) is a skeletal muscle relaxant and a central nervous system (CNS) depressant.

Cyclobenzaprine hydrochloride (MK130 hydrochloride) is a skeletal muscle relaxant and a central nervous system (CNS) depressant.

Specifications

Chemical Name or Material Cyclobenzaprine hydrochloride
Physical Form Solid
CAS 6202-23-9
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C20H22ClN
For Use With (Application) Neuroscience-Neuromodulation
Synonym MK130 hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (320.67 mM) ∣DMSO : 50 mg/mL (160.33 mM; Need ultrasonic)
SMILES CN(C)CC/C=C1C2=CC=CC=C2C=CC3=CC=CC=C\13.Cl
Molecular Weight (g/mol) 311.85
Formula Weight 311.85
Percent Purity 99.91%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.