missing translation for 'onlineSavingsMsg'
Learn More

Cyclobenzaprine Hydrochloride, TRC

Product Code. 30040474
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040474 500 mg 500mg
18012350 100 mg 100mg
18081150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040474 Supplier Toronto Research Chemicals Supplier No. TRCC987750500MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6202-23-9
Molecular Formula C20 H21 N . Cl H
Molecular Weight (g/mol) 311.85
Synonym Cyclobenzaprine Hydrochloride, 3-(5H-Dibenzo[a,d][7]annulen-5-ylidene)-N,N-dimethylpropan-1-amine Hydrochloride, Nortriptyline Hydrochloride Imp. E (EP) as Hydrochloride, Amitriptyline Hydrochloride Imp. B (EP) as Hydrochloride, 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (1:1), 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-dimethyl-, hydrochloride (9CI), 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, N,N-dimethyl-, hydrochloride (6CI,7CI,8CI), 10,11δ-Amitriptyline hydrochloride, Benzamin, Flexeril, Flexiban, Lisseril, MK 130 hydrochloride, Miosan, Multi-Relax, Proheptatriene hydrochloride, Tensodox, Yurelax
IUPAC Name 3-(dibenzo[2,1-b:1',2'-e][7]annulen-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCC=C1c2ccccc2C=Cc3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H21N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-14H,7,15H2,1-2H3;1H
Formula Weight 311.1441
Percent Purity >95%
Chemical Name or Material Cyclobenzaprine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.