missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone 5000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30326410
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326410 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326410 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010400ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclohexanone
CAS 108-94-1
Analyte or Component Names Cyclohexanone
Shipping Condition Ice pack (-20°C)
Molecular Formula C6 H10 O
Concentration 5000 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclohexanone, Anon, Anone, C 0489, Hexanon, Hytrol O, NSC 5711, Nadone, Pimelic ketone, Pimelin ketone, Sextone
InChI Formula InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2
SMILES O=C1CCCCC1
IUPAC Name cyclohexanone
Solvent or Matrix Methanol
Molecular Weight (g/mol) 98.14
Quantity 1 mL
Formula Weight 98.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.