missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cycloheximide, ultrapure, 95%

Product Code. 15458449
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458449 Supplier Thermo Scientific Chemicals Supplier No. J66901.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 66-81-9 | C15H23NO4 | 281.35 g/mol

A protein synthesis inhibitor in eukaryotic cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Type Cycloheximide
Physical Form Powder
CAS 66-81-9
Melting Point 111°C to 116°C
Color Cream to White
Quantity 1 g
Flash Point >110°C (230°F)
Molecular Formula C15H23NO4
MDL Number MFCD00082346
UN Number UN2811
Beilstein 88868
Boiling Point 492°C
Merck Index 14,2728
Synonym 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
Solubility Information Soluble in DMSO, acetone, ethanol and methanol; Slightly soluble in water with sonication
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Molecular Weight (g/mol) 281.35
PubChem CID 6197
ChEBI CHEBI:27641
Formula Weight 281.35
Percent Purity 95%
Chemical Name or Material Cycloheximide, Ultrapure
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cycloheximide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H300-Fatal if swallowed.
  • H341-Suspected of causing genetic defects.
  • H360D-May damage the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Cycloheximide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.