missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%

Product Code. 10350984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10350984 25 g 25g
10385690 100 g 100g
10289393 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10350984 Supplier Thermo Scientific Acros Supplier No. 111440250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 502-49-8 | C8H14O | 126.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-49-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
MDL Number MFCD00001754
InChI Key IIRFCWANHMSDCG-UHFFFAOYSA-N
Synonym cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
PubChem CID 10403
IUPAC Name cyclooctanone
SMILES C1CCCC(=O)CCC1

Specifications

Melting Point 39.0°C to 44.0°C
Color Colorless to White
Density 0.9500g/mL
Boiling Point 195.0°C to 197.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C8H14(=O)
Quantity 25 g
Beilstein 07, 21
Specific Gravity 0.95
Solubility Information Solubility in water: 15g/l (20°C). Other solubilities: soluble in alcohol,acetone and benzene
Formula Weight 126.2
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Cyclooctanone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclooctanone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.