missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclophilin B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00 € - 401.00 €
Specifications
| Antigen | Cyclophilin B |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml, Knockout Validated |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110512
|
Novus Biologicals
NBP2-24770SS |
0.025 mg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768443
|
Novus Biologicals
NBP2-24770 |
0.1 mg |
401.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclophilin B Polyclonal specifically detects Cyclophilin B in Human, Mouse, Porcine, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Knockout Validated.Specifications
| Cyclophilin B | |
| Western Blot 0.5-2 ug/ml, Knockout Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Pig, Bovine, Canine, Equine, Primate | |
| P23284 | |
| 5479 | |
| The portions of amino acids range between 100-228 of human Cyclophilin B was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Loading Controls | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP | |
| PPIB | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog and bovine, and 84% in chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title