missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide hydrate, MedChemExpress

Product Code. 16467296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467296 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16467296 Supplier Medchem Express Supplier No. HY17420A
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cyclophosphamide hydrate is a synthetic alkylating agent chemically related to the nitrogen mustards with antineoplastic and immunosuppressive activities.

Cyclophosphamide hydrate is a synthetic alkylating agent chemically related to the nitrogen mustards with antineoplastic and immunosuppressive activities.

Specifications

Chemical Name or Material Cyclophosphamide hydrate
Physical Form Solid
CAS 6055-19-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H17Cl2N2O3P
For Use With (Application) Cancer-programmed cell death
Synonym Cyclophosphamide monohydrate
Solubility Information H2O : ≥ 50 mg/mL (179.15 mM) ∣DMSO : ≥ 38 mg/mL (136.15 mM)
SMILES ClCCN(CCCl)P1(OCCCN1)=O.[H]O[H]
Molecular Weight (g/mol) 279.1
Formula Weight 279.1
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.