missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxyl Carnitine Chloride, TRC

Product Code. 30137408
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137408 10 mg 10mg
30007606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137408 Supplier Toronto Research Chemicals Supplier No. TRCC17915710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11 H20 N O4 . Cl
Molecular Weight (g/mol) 265.73
IUPAC Name [3-carboxy-2-(cyclopropanecarbonyloxy)propyl]-trimethyl-ammonium;chloride
SMILES [Cl-].C[N+](C)(C)CC(CC(=O)O)OC(=O)C1CC1

Specifications

Analyte or Component Names Cyclopropanecarboxyl Carnitine Chloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C11H19NO4.ClH/c1-12(2,3)7-9(6-10(13)14)16-11(15)8-4-5-8;/h8-9H,4-7H2,1-3H3;1H
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 265.1081
Chemical Name or Material Cyclopropanecarboxyl Carnitine Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.